missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9,10-Bis(6-methoxy-2-naphthyl)anthracene 98.0+%, TCI America™
$229.10 - $685.36
Chemical Identifiers
| CAS | 235099-48-6 |
|---|---|
| Molecular Formula | C36H26O2 |
| Molecular Weight (g/mol) | 490.602 |
| MDL Number | MFCD12022382 |
| InChI Key | VEVBQRGODPIBNJ-UHFFFAOYSA-N |
| PubChem CID | 18710993 |
| IUPAC Name | 9,10-bis(6-methoxynaphthalen-2-yl)anthracene |
| SMILES | COC1=CC2=C(C=C1)C=C(C=C2)C3=C4C=CC=CC4=C(C5=CC=CC=C53)C6=CC7=C(C=C6)C=C(C=C7)OC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
B5046200MG
|
TCI America
B5046200MG |
200 mg |
Each for $229.10
|
|
|||||
|
B50461G
|
TCI America
B50461G |
1 g |
Each for $685.36
|
|
|||||
Chemical Identifiers
| 235099-48-6 | |
| 490.602 | |
| VEVBQRGODPIBNJ-UHFFFAOYSA-N | |
| 9,10-bis(6-methoxynaphthalen-2-yl)anthracene |
| C36H26O2 | |
| MFCD12022382 | |
| 18710993 | |
| COC1=CC2=C(C=C1)C=C(C=C2)C3=C4C=CC=CC4=C(C5=CC=CC=C53)C6=CC7=C(C=C6)C=C(C=C7)OC |
Specifications
| 235099-48-6 | |
| C36H26O2 | |
| 200 mg | |
| COC1=CC2=C(C=C1)C=C(C=C2)C3=C4C=CC=CC4=C(C5=CC=CC=C53)C6=CC7=C(C=C6)C=C(C=C7)OC | |
| 490.602 | |
| 490.60 | |
| Crystalline Powder |
| White-Yellow | |
| MFCD12022382 | |
| VEVBQRGODPIBNJ-UHFFFAOYSA-N | |
| 9,10-bis(6-methoxynaphthalen-2-yl)anthracene | |
| 18710993 | |
| ≥98.0% (HPLC) | |
| 9,10-Bis(6-methoxy-2-naphthyl)anthracene |
Safety and Handling
TSCA : No