missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9,10-Bis(6-methoxy-2-naphthyl)anthracene 98.0+%, TCI America™
Specifications
Specifications
| Color | White-Yellow |
| Quantity | 200 mg |
| Formula Weight | 490.60 |
| Percent Purity | ≥98.0% (HPLC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 9,10-Bis(6-methoxy-2-naphthyl)anthracene |
Safety and Handling
| TSCA | No |