missing translation for 'onlineSavingsMsg'
Learn More

9,10-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)anthracene 98.0+%, TCI America™

Catalog No. B45625G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B45625G 5 g
B456225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B45625G Supplier TCI America Supplier No. B45625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 863992-56-7
Molecular Formula C26H32B2O4
Molecular Weight (g/mol) 430.16
MDL Number MFCD16294538
InChI Key ZLXSWNVYGSZXOP-UHFFFAOYSA-N
Synonym 9,10-anthracenediboronic acid bis pinacol ester, 9,10-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl anthracene, 4,4,5,5-tetramethyl-2-10-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl anthracen-9-yl-1,3,2-dioxaborolane, amtb237, anthracene-9,10-diboronic acid bis pinacol ester, 9,10-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolanyl anthracene, 9,10-bis 4,4,5,5-tetramethyl 1.3.2 dioxaborolan-2-yl anthracene, 9,10-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl anthracene, 1,3,2-dioxaborolane, 2,2'-9,10-anthracenediyl bis 4,4,5,5-tetramethyl
PubChem CID 57415691
IUPAC Name 4,4,5,5-tetramethyl-2-[10-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)anthracen-9-yl]-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)C1=C2C=CC=CC2=C(B2OC(C)(C)C(C)(C)O2)C2=CC=CC=C12

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 430.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9,10-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)anthracene