missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9-(1-Naphthyl)anthracene 98.0+%, TCI America™
Supplier: TCI America N09945G
Specifications
| 9-(1-Naphthyl)anthracene | |
| 165°C | |
| C24H16 | |
| 5 g | |
| C1=CC2=CC3=CC=CC=C3C(C3=CC=CC4=CC=CC=C34)=C2C=C1 | |
| 304.39 | |
| 304.39 | |
| Crystalline Powder |
| 7424-70-6 | |
| White-Yellow | |
| MFCD19689379 | |
| YPNZWHZIYLWEDR-UHFFFAOYSA-N | |
| 9-(naphthalen-1-yl)anthracene | |
| 18510032 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 7424-70-6 | |
| 304.39 | |
| YPNZWHZIYLWEDR-UHFFFAOYSA-N | |
| 9-(naphthalen-1-yl)anthracene |
| C24H16 | |
| MFCD19689379 | |
| 18510032 | |
| C1=CC2=CC3=CC=CC=C3C(C3=CC=CC4=CC=CC=C34)=C2C=C1 |
Safety and Handling
TSCA : No