missing translation for 'onlineSavingsMsg'
Learn More

9-(1-Naphthyl)anthracene 98.0+%, TCI America™

Catalog No. p-7134071
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N09941G 1 g
N09945G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N09941G Supplier TCI America Supplier No. N09941G

Chemical Identifiers

CAS 7424-70-6
Molecular Formula C24H16
Molecular Weight (g/mol) 304.39
MDL Number MFCD19689379
InChI Key YPNZWHZIYLWEDR-UHFFFAOYSA-N
PubChem CID 18510032
IUPAC Name 9-(naphthalen-1-yl)anthracene
SMILES C1=CC2=CC3=CC=CC=C3C(C3=CC=CC4=CC=CC=C34)=C2C=C1

Specifications

Melting Point 165°C
Color White-Yellow
Quantity 1 g
Formula Weight 304.39
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9-(1-Naphthyl)anthracene