missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9-([1,1'-Biphenyl]-4-yl)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole 97.0+%, TCI America™
Specifications
Specifications
| Melting Point | 231°C |
| Color | White-Yellow |
| Quantity | 1 g |
| Synonym | 9-(4-Biphenylyl)carbazole-3-boronic Acid Pinacol Ester, 2-[9-([1,1′C-Biphenyl]-4-yl)-9H-carbazol-3-yl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane |
| Formula Weight | 445.37 |
| Percent Purity | ≥97.0% (HPLC,N) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 9-([1,1′-Biphenyl]-4-yl)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole |
Safety and Handling
| TSCA | No |