missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ 81 Well Tube Storage Box

The 81 well microtube storage box has a compact footprint for microtube and cryogenic vial storage

Supplier:  Fisherbrand™ 120034F2

 View more versions of this product

Catalog No. 14127119



Only null left

Description

Description

  • Made of vibrantly colored polypropylene which allows for visibility inside the box and color-coding identification
  • Tube capacity is indicated on the outside the box within a distinctive wave design
  • Quick visual identification with uniform wells and alphanumeric identification along with a frosted labeling area
  • Easy open and secure lid fitted with friction fit design
  • Stackable, saving valuable space in the lab
  • Can be used in temperatures as low as -80°C
Specifications

Chemical Identifiers

Hinged Lid
Square
Non-sterile
-80°C
Polypropylene
Cryo Storage Box
9 x 9
5.1 x 5.1 x 1.9 in.

Specifications

Hinged Lid
Polypropylene
Cryo Storage Box
9 x 9
5.1 x 5.1 x 1.9 in.
Natural
Square
Non-sterile
-80°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions