Learn More
Cole-Parmer™ Adapters and Clamps for High-Energy Mixer/Mill™ Grinders
Description
- Suitable for use with the Cole-Parmer™ Freezer/Mill™ Cryogenic Grinders
- Optimize performance of your Cole-Parmer™ Freezer/Mill™ grinder with a wide range of adapters and clamps.
Specifications
Specifications
| Product Type | Clamps for slurry grinding |
| For Use With (Application) | Ideal for High Throughput Applications That Involve Processing Multiple Smaller Samples |
| Holds | Alumina Ceramic Grinding Vial Set (41005-86) |