Learn More
Cole-Parmer™ Adapters and Clamps for High-Energy Mixer/Mill™ Grinders
Catalog No. 8011
Change view
Click to view available options
Product Type:
Clamps for slurry grinding
Multiple Sample Adapter
Vial Clamp Adapter
Holds:
4 x 0.75 in. Vials
7 x 0.5 in. dia. Vials
Alumina Ceramic Grinding Vial Set (41005-86)
Vial Adapters (04576-65 and 04576-66)
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Holds
8011 Multiple Sample Adapter 4 x 0.75 in. Vials
8010 Multiple Sample Adapter 7 x 0.5 in. dia. Vials
8015 Clamps for slurry grinding Alumina Ceramic Grinding Vial Set (41005-86)
8012 Vial Clamp Adapter Vial Adapters (04576-65 and 04576-66)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 8011 Supplier Cole-Parmer™ Supplier No. 8011
Use adapters to run multiple samples or to hold vial securely in place during grinding.
  • Suitable for use with the Cole-Parmer™ Freezer/Mill™ Cryogenic Grinders
  • Optimize performance of your Cole-Parmer™ Freezer/Mill™ grinder with a wide range of adapters and clamps.
Specifications
Product Type Multiple Sample Adapter
For Use With (Application) Ideal for High Throughput Applications That Involve Processing Multiple Smaller Samples
For Use With (Equipment) 8000 D and 8000 M Mixer or Mill
Holds 4 x 0.75 in. Vials