missing translation for 'onlineSavingsMsg'
Learn More

Agilent BioTek 800 TS Absorbance Reader, IVD
GREENER_CHOICE

Catalog No. BT800TUVISI
Change view
Click to view available options
Filters:
340, 405, 450, 490, 630 nm
405, 450, 490, 630 nm
Includes:
6- to 384-well Terasaki Plates
6- to 96-well Plates, Shaking
6- to 96-well Plates, Shaking, Incubation
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Filters Includes
BT800TUVISI 340, 405, 450, 490, 630 nm 6- to 96-well Plates, Shaking, Incubation
BT800TSSI 405, 450, 490, 630 nm 6- to 96-well Plates, Shaking
BT800TSISI 405, 450, 490, 630 nm 6- to 96-well Plates, Shaking, Incubation
BT800TSNBSI 405, 450, 490, 630 nm 6- to 384-well Terasaki Plates
BT800TSUVSI 340, 405, 450, 490, 630 nm 6- to 96-well Plates, Shaking
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. BT800TUVISI Supplier Agilent BioTek Supplier No. 800TSUVISI

missing translation for 'validMainframeMsg'

The BioTek 800 TS absorbance reader is an affordable, high-quality microplate reader for assays in 6- to 384-well formats. For In Vitro Diagnostic Use.

The BioTek 800 TS absorbance reader is an affordable, high-quality microplate reader for assays in 6- to 384-well formats. Applications include ELISA, protein quantification, enzyme kinetics and cell-based assays

The color touch screen provides a visual user interface, making programming fast and intuitive. Expanded functionality and data analysis is available with Gen5 software. The 800 TS partners with the 50 TS washer, making a compact, versatile, and affordable system to automate many application workflows.

Specifications

Product Type Microplate Reader
Detector Type Photodiode
Interface USB
Light Source Tungsten Halogen Lamp
Reading Options Endpoint, Kinetic
Shaking Linear (except NB configurations)
Software Type Gen5 Data Analysis Software, Gen5 Secure for 21 CFR Part 11 compliance (option)
Bandwidth 10 nm
Certifications/Compliance CE and TUV marked. RoHS compliant.
Filters 340, 405, 450, 490, 630 nm
For Use With (Application) Measuring absorbance in microplate-based assays
Includes 6- to 96-well Plates, Shaking, Incubation
Model 800TSUVI
Voltage 100/240 V
Wavelength Range 340 to 750 nm
Weight (English) 18.5 lb.
Weight (Metric) 8 kg
Show More Show Less