Learn More
Agilent BioTek 800 TS Absorbance Reader, IVD

The BioTek 800 TS absorbance reader is an affordable, high-quality microplate reader for assays in 6- to 384-well formats. For In Vitro Diagnostic Use.
Supplier: Agilent BioTek 800TSUVSI
Description
The BioTek 800 TS absorbance reader is an affordable, high-quality microplate reader for assays in 6- to 384-well formats. Applications include ELISA, protein quantification, enzyme kinetics and cell-based assays
The color touch screen provides a visual user interface, making programming fast and intuitive. Expanded functionality and data analysis is available with Gen5 software. The 800 TS partners with the 50 TS washer, making a compact, versatile, and affordable system to automate many application workflows.
Chemical Identifiers
| Microplate Reader | |
| USB | |
| Endpoint, Kinetic, Well-area Scanning | |
| Gen5 Data Analysis Software, Gen5 Secure for 21 CFR Part 11 compliance (option) | |
| CE and TUV marked. RoHS compliant. | |
| 100/240 V | |
| 8 kg |
| Photodiode | |
| Tungsten Halogen Lamp | |
| Linear (except NB configurations) | |
| 10 nm | |
| Measuring absorbance in microplate-based assays | |
| 18.5 lb. |
Specifications
| Microplate Reader | |
| USB | |
| Endpoint, Kinetic, Well-area Scanning | |
| Gen5 Data Analysis Software, Gen5 Secure for 21 CFR Part 11 compliance (option) | |
| CE and TUV marked. RoHS compliant. | |
| Measuring absorbance in microplate-based assays | |
| 800TSUV | |
| 340 to 750 nm | |
| 8 kg |
| Photodiode | |
| Tungsten Halogen Lamp | |
| Linear (except NB configurations) | |
| 10 nm | |
| 340, 405, 450, 490, 630 nm | |
| 6- to 96-well Plates, Shaking | |
| 100/240 V | |
| 18.5 lb. |