Learn More
Agilent BioTek 800 TS Absorbance Reader
GREENER_CHOICE
The BioTek 800 TS absorbance reader is an affordable, high-quality microplate reader for assays in 6- to 384-well formats. For Research Use Only. Not for use in diagnostic procedures.
Supplier:  Agilent BioTek 800TSUVSN
 View more versions of this product
Catalog No. BT800TSUV
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
The BioTek 800 TS absorbance reader is an affordable, high-quality microplate reader for assays in 6- to 384-well formats. Applications include ELISA, protein quantification, enzyme kinetics and cell-based assays
The color touch screen provides a visual user interface, making programming fast and intuitive. Expanded functionality and data analysis is available with Gen5 software. The 800 TS partners with the 50 TS washer, making a compact, versatile, and affordable system to automate many application workflows.
Warranty
12 months
Specifications
Chemical Identifiers
Microplate Reader
USB Type B, USB Type A
Endpoint, Kinetic, Well-area Scanning
Gen5 Data Analysis Software, Gen5 Secure for 21 CFR Part 11 compliance (option)
CE and TUV marked. RoHS compliant.
100/240 V
8 kg
Photodiode
Tungsten Halogen Lamp
Linear (except NB configurations)
10 nm
Measuring absorbance in microplate-based assays
18.5 lb.
Specifications
Microplate Reader
USB Type B, USB Type A
Endpoint, Kinetic, Well-area Scanning
Gen5 Data Analysis Software, Gen5 Secure for 21 CFR Part 11 compliance (option)
CE and TUV marked. RoHS compliant.
Measuring absorbance in microplate-based assays
800TSUV
340 to 750 nm
8 kg
Photodiode
Tungsten Halogen Lamp
Linear (except NB configurations)
10 nm
340, 405, 450, 490, 630 nm
6- to 96-well Plates, Shaking
100/240 V
18.5 lb.
Product Suggestions
Product Suggestions