missing translation for 'onlineSavingsMsg'
Learn More

8-(Trifluoromethyl)-4-quinolinol 98.0+%, TCI America™

Catalog No. T31601G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T31601G 1 g
T31605G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T31601G Supplier TCI America Supplier No. T31601G

Chemical Identifiers

CAS 23779-96-6
Molecular Formula C10H6F3NO
Molecular Weight (g/mol) 213.159
MDL Number MFCD00134577
InChI Key UDRWADJLLWWJOE-UHFFFAOYSA-N
Synonym 8-trifluoromethyl quinolin-4-ol, 4-hydroxy-8-trifluoromethyl quinoline, 8-trifluoromethyl-4-quinolinol, 4-hydroxy-8-trifluoromethylquinoline, 8-trifluoromethyl-1h-quinolin-4-one, 8-trifluoromethyl quinolin-4 1h-one, 8-trifluoromethyl-quinolin-4-ol, 4-quinolinol,8-trifluoromethyl, 4-quinolinol, 8-trifluoromethyl
PubChem CID 90261
IUPAC Name 8-(trifluoromethyl)-1H-quinolin-4-one
SMILES C1=CC2=C(C(=C1)C(F)(F)F)NC=CC2=O

Specifications

Melting Point 174°C
Quantity 1 g
Formula Weight 213.16
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 8-(Trifluoromethyl)-4-quinolinol