CAS RN 23779-96-6
IUPAC Name:
8-(trifluoromethyl)-1,4-dihydroquinolin-4-one
Synonyms:
8-(trifluoromethyl)-1H-quinolin-4-one
Molecular Weight (g/mol):
213.16
Molecular Formula:
C10H6F3NO
InChi Key:
UDRWADJLLWWJOE-UHFFFAOYSA-N
SMILES:
FC(F)(F)C1=C2NC=CC(=O)C2=CC=C1
1
–
1
of
1
results
1
8-(Trifluoromethyl)-4-quinolinol 98.0+%, TCI America™
CAS: 23779-96-6 Molecular Formula: C10H6F3NO Molecular Weight (g/mol): 213.159 MDL Number: MFCD00134577 InChI Key: UDRWADJLLWWJOE-UHFFFAOYSA-N Synonym: 8-trifluoromethyl quinolin-4-ol,4-hydroxy-8-trifluoromethyl quinoline,8-trifluoromethyl-4-quinolinol,4-hydroxy-8-trifluoromethylquinoline,8-trifluoromethyl-1h-quinolin-4-one,8-trifluoromethyl quinolin-4 1h-one,8-trifluoromethyl-quinolin-4-ol,4-quinolinol,8-trifluoromethyl,4-quinolinol, 8-trifluoromethyl PubChem CID: 90261 IUPAC Name: 8-(trifluoromethyl)-1H-quinolin-4-one SMILES: C1=CC2=C(C(=C1)C(F)(F)F)NC=CC2=O