missing translation for 'onlineSavingsMsg'
Learn More

8-Quinolinol 99.0+%, TCI America™

Catalog No. p-7130477
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H030525G 25 g
H0305500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H030525G Supplier TCI America Supplier No. H030525G
Only null left

Chemical Identifiers

CAS 148-24-3
Molecular Formula C9H7NO
Molecular Weight (g/mol) 145.16
MDL Number MFCD00006807
InChI Key MCJGNVYPOGVAJF-UHFFFAOYSA-N
Synonym 8-hydroxyquinoline, 8-quinolinol, oxyquinoline, oxine, quinophenol, 8-quinol, 8-oxyquinoline, phenopyridine, oxychinolin, bioquin
PubChem CID 1923
ChEBI CHEBI:48981
IUPAC Name quinolin-8-ol
SMILES OC1=C2N=CC=CC2=CC=C1

Specifications

Melting Point 75°C
Color White-Yellow
Boiling Point 126°C
Quantity 25 g
UN Number 3077
Formula Weight 145.16
Percent Purity ≥99.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 8-Quinolinol
EINECSNumber (5)-0804
RTECSNumber VC4200000
TSCA Yes