Learn More
Eisco™ 8 Piece Tuning Fork Sets with Case
8 piece steel tuning forks with Scientific Pitch, C4 = 256Hz are designed for sound wave lessons and exploration
Supplier:  Eisco™ PH0738C
 View more versions of this product
Catalog No. S27733
Only null left
Explore available promotions
Description
Description
  • Supplied with either a plastic, or wooden case for long term storage
  • Tuning forks are nickel plated with plain shanks
  • Frequency is marked on each tuning fork
  • Includes 1 of each: C (256), D (288), E (320), F (341), G (384), A (426), B (480), C (512)
Specifications
Specifications
Tuning Fork Set, 8 Piece
Steel Tuning Fork
Plastic Case
Scientific Pitch (C4 = 256 Hz)
Sound Wave Lessons and Exploration
Product Suggestions
Product Suggestions