missing translation for 'onlineSavingsMsg'
Learn More

8-Oxa-3-azabicyclo[3.2.1]octane hydrochloride, 97%

Catalog No. aah63825md
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH63825MD 250 mg
AAH6382503 1 g
AAH6382506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH63825MD Supplier Thermo Scientific Chemicals Supplier No. H63825MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 54745-74-3 | C6H12ClNO | 149.62 g/mol

8-Oxa-3-azabicyclo[3.2.1]octane hydrochloride is employed as a important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
8-Oxa-3-azabicyclo[3.2.1]octane hydrochloride is employed as a important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 54745-74-3
Molecular Formula C6H12ClNO
Molecular Weight (g/mol) 149.62
MDL Number MFCD17926456,MFCD09800611
InChI Key XADOTNAXKKFKDY-UHFFFAOYNA-N
Synonym 8-oxa-3-azabicyclo 3.2.1 octane hydrochloride, 8-oxa-3-azabicyclo 3,2,1 octane hydrochloride, 8-oxa-3-azabicyclo 3.2.1 octane hcl, 8-oxa-3-azabicyclo 3.2.1 octanehydrochloride, 8-oxa-3-azabicyclo 3.2.1 octane, hydrochloride, 8-oxa-3-aza-bicyclo 3.2.1 octane hydrochloride, 8-oxa-3-azabicyclooctane hydrochloride, 8-oxa-3-azabicyclo 3.2.1 octane-hcl, 8-oxa-3-aza-bicyclo 3.2.1 octane hcl
PubChem CID 21983536
SMILES Cl.C1CC2CNCC1O2

Specifications

Quantity 250 mg
IUPAC Name 8-oxa-3-azabicyclo[3.2.1]octane;hydrochloride
Formula Weight 149.62
Percent Purity 97%
Chemical Name or Material 8-Oxa-3-azabicyclo[3.2.1]octane hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only