CAS RN 54745-74-3
IUPAC Name:
8-oxa-3-azabicyclo[3.2.1]octane hydrochloride
Synonyms:
8-oxa-3-azabicyclo[3.2.1]octane hydrochloride
Molecular Weight (g/mol):
149.62
Molecular Formula:
C6H12ClNO
InChi Key:
XADOTNAXKKFKDY-UHFFFAOYNA-N
SMILES:
Cl.C1CC2CNCC1O2
1
–
1
of
1
results
1
Promotions Available
8-Oxa-3-azabicyclo[3.2.1]octane hydrochloride, 97%
8-Oxa-3-azabicyclo[3.2.1]octane hydrochloride, 97%
CAS: 54745-74-3 Molecular Formula: C6H12ClNO Molecular Weight (g/mol): 149.62 MDL Number: MFCD17926456,MFCD09800611 InChI Key: XADOTNAXKKFKDY-UHFFFAOYNA-N Synonym: 8-oxa-3-azabicyclo 3.2.1 octane hydrochloride,8-oxa-3-azabicyclo 3,2,1 octane hydrochloride,8-oxa-3-azabicyclo 3.2.1 octane hcl,8-oxa-3-azabicyclo 3.2.1 octanehydrochloride,8-oxa-3-azabicyclo 3.2.1 octane, hydrochloride,8-oxa-3-aza-bicyclo 3.2.1 octane hydrochloride,8-oxa-3-azabicyclooctane hydrochloride,8-oxa-3-azabicyclo 3.2.1 octane-hcl,8-oxa-3-aza-bicyclo 3.2.1 octane hcl PubChem CID: 21983536 SMILES: Cl.C1CC2CNCC1O2