missing translation for 'onlineSavingsMsg'
Learn More
Learn More
8-Nonenyl Acetate 98.0+%, TCI America™
Supplier: TCI America A138425ML
Specifications
| 8-Nonenyl Acetate | |
| Yellow | |
| C11H20O2 | |
| 25 mL | |
| HUXQMCFHKGATCP-UHFFFAOYSA-N | |
| non-8-enyl acetate | |
| 15673852 | |
| ≥98.0% (GC) |
| 13038-22-7 | |
| 155°C | |
| MFCD00671492 | |
| 9-Acetoxy-1-nonene, Acetic Acid 8-Nonenyl Ester | |
| CC(=O)OCCCCCCCC=C | |
| 184.279 | |
| 184.28 | |
| Liquid |
Chemical Identifiers
| 13038-22-7 | |
| 184.279 | |
| HUXQMCFHKGATCP-UHFFFAOYSA-N | |
| 15673852 | |
| CC(=O)OCCCCCCCC=C |
| C11H20O2 | |
| MFCD00671492 | |
| 9-Acetoxy-1-nonene, Acetic Acid 8-Nonenyl Ester | |
| non-8-enyl acetate |
Safety and Handling
EINECSNumber : (2)-0762
TSCA : No