missing translation for 'onlineSavingsMsg'
Learn More

8-Nonenyl Acetate 98.0+%, TCI America™

Catalog No. p-7117400
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A13845ML 5 mL
A138425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A13845ML Supplier TCI America Supplier No. A13845ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13038-22-7
Molecular Formula C11H20O2
Molecular Weight (g/mol) 184.279
MDL Number MFCD00671492
InChI Key HUXQMCFHKGATCP-UHFFFAOYSA-N
Synonym 9-Acetoxy-1-nonene, Acetic Acid 8-Nonenyl Ester
PubChem CID 15673852
IUPAC Name non-8-enyl acetate
SMILES CC(=O)OCCCCCCCC=C

Specifications

Color Yellow
Boiling Point 155°C
Quantity 5 mL
Formula Weight 184.28
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 8-Nonenyl Acetate
EINECSNumber (2)-0762
TSCA No