missing translation for 'onlineSavingsMsg'
Learn More

8-Nonen-1-ol 98.0+%, TCI America™

Supplier:  TCI America N065025G

 View more versions of this product

Catalog No. N065025G


Only null left

Description

Description

Specifications

Specifications

8-Nonen-1-ol
Yellow
C9H18O
25 g
C=CCCCCCCCO
142.242
142.24
Liquid
13038-21-6
134°C
MFCD00798062
FKGFCVJJLGSFSB-UHFFFAOYSA-N
non-8-en-1-ol
11094728
≥98.0% (GC)

Chemical Identifiers

13038-21-6
142.242
FKGFCVJJLGSFSB-UHFFFAOYSA-N
non-8-en-1-ol
C9H18O
MFCD00798062
11094728
C=CCCCCCCCO
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions