missing translation for 'onlineSavingsMsg'
Learn More

8-Nonen-1-ol 98.0+%, TCI America™

Catalog No. p-7133909
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N06505G 5 g
N065025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N06505G Supplier TCI America Supplier No. N06505G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13038-21-6
Molecular Formula C9H18O
Molecular Weight (g/mol) 142.242
MDL Number MFCD00798062
InChI Key FKGFCVJJLGSFSB-UHFFFAOYSA-N
PubChem CID 11094728
IUPAC Name non-8-en-1-ol
SMILES C=CCCCCCCCO

Specifications

Color Yellow
Boiling Point 134°C
Quantity 5 g
Formula Weight 142.24
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 8-Nonen-1-ol