missing translation for 'onlineSavingsMsg'
Learn More

8-Nitro-7-quinolinecarboxaldehyde 98.0+%, TCI America™

Catalog No. N08521G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N08521G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N08521G Supplier TCI America Supplier No. N08521G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 101327-87-1
Molecular Formula C10H6N2O3
Molecular Weight (g/mol) 202.169
InChI Key CGYVBVWBGRTQJQ-UHFFFAOYSA-N
Synonym 7-Formyl-8-nitroquinoline
PubChem CID 11769554
IUPAC Name 8-nitroquinoline-7-carbaldehyde
SMILES C1=CC2=C(C(=C(C=C2)C=O)[N+](=O)[O-])N=C1

Specifications

Melting Point 180°C
Quantity 1 g
Formula Weight 202.17
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 8-Nitro-7-quinolinecarboxaldehyde