accessibility menu, dialog, popup

UserName

ONOON

CAS RN 101327-87-1

IUPAC Name: 8-nitroquinoline-7-carbaldehyde
Synonyms: 8-nitroquinoline-7-carbaldehyde
Molecular Weight (g/mol): 202.17
Molecular Formula: C10H6N2O3
InChi Key: CGYVBVWBGRTQJQ-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=C(C=O)C=CC2=CC=CN=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results