missing translation for 'onlineSavingsMsg'
Learn More

8-Methylquinoline 99.0+%, TCI America™

Supplier:  TCI America M041925G

 View more versions of this product

Catalog No. M041925G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

8-Methylquinoline
-80°C
248°C
MFCD00006810
quinoline, 8-methyl, 8-methyl-quinoline, ccris 408, 8-methyl, o-toluquinoline, 8-metyl quinoline, 8-methyl quinoline, pubchem5806, acmc-209mpf, dsstox_cid_888
CC1=C2N=CC=CC2=CC=C1
143.19
CHEBI:48984
≥99.0% (GC)
611-32-5
Yellow
C10H9N
25 g
JRLTTZUODKEYDH-UHFFFAOYSA-N
8-methylquinoline
11910
143.19
Liquid

Chemical Identifiers

611-32-5
143.19
JRLTTZUODKEYDH-UHFFFAOYSA-N
11910
8-methylquinoline
C10H9N
MFCD00006810
quinoline, 8-methyl, 8-methyl-quinoline, ccris 408, 8-methyl, o-toluquinoline, 8-metyl quinoline, 8-methyl quinoline, pubchem5806, acmc-209mpf, dsstox_cid_888
CHEBI:48984
CC1=C2N=CC=CC2=CC=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (5)-0796

RTECSNumber : VC0562000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions