missing translation for 'onlineSavingsMsg'
Learn More

8-Methylquinoline 99.0+%, TCI America™

Catalog No. p-7132159
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M04195G 5 g
M041925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M04195G Supplier TCI America Supplier No. M04195G
Only null left

Chemical Identifiers

CAS 611-32-5
Molecular Formula C10H9N
Molecular Weight (g/mol) 143.19
MDL Number MFCD00006810
InChI Key JRLTTZUODKEYDH-UHFFFAOYSA-N
Synonym quinoline, 8-methyl, 8-methyl-quinoline, ccris 408, 8-methyl, o-toluquinoline, 8-metyl quinoline, 8-methyl quinoline, pubchem5806, acmc-209mpf, dsstox_cid_888
PubChem CID 11910
ChEBI CHEBI:48984
IUPAC Name 8-methylquinoline
SMILES CC1=C2N=CC=CC2=CC=C1

Specifications

Melting Point -80°C
Color Yellow
Boiling Point 248°C
Quantity 5 g
Formula Weight 143.19
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 8-Methylquinoline
EINECSNumber (5)-0796
RTECSNumber VC0562000
TSCA Yes