missing translation for 'onlineSavingsMsg'
Learn More

8-Methoxypsoralen, MilliporeSigma™ Supelco™

Catalog No. 111012190
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012190 1 g
111012191 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111012190 Supplier Merck Emd Millipore Supplier No. M35011G
Only null left
Food and Beverages

Specifications

Packaging Glass Bottle
CAS 298-81-7
Physical Form Crystals
Linear Formula C12H8O4
MDL Number MFCD00005009
Melting Point 148°C to 150°C
Synonym 8-MOP; 9-Methoxyfuro[3,2-g][1 ]benzopyran-7-one; Ammoidin; Methoxsalen; Xanthotoxin
Solubility Soluble in acetic acid
Type Analytical Standard
Quantity 1 g
Formula Weight 216.19
Grade Analytical Standard
Show More Show Less