missing translation for 'onlineSavingsMsg'
Learn More

8-Methoxyjulolidine, 95%

Catalog No. AAH3410009
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3410009 10 g
AAH3410003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3410009 Supplier Thermo Scientific Chemicals Supplier No. H3410009
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 63468-83-7 | C13H17NO | 203.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 63468-83-7
Molecular Formula C13H17NO
Molecular Weight (g/mol) 203.29
MDL Number MFCD00037096
InChI Key ZSKPEBROOQBZNW-UHFFFAOYSA-N
Synonym 8-methoxyjulolidine, 8-methoxy-1,2,3,5,6,7-hexahydropyrido 3,2,1-ij quinoline, enamine_005788, 8-methoxy-1-azatricyclo 7.3.1.0?, 1 3 trideca-5,7,9 13-triene
PubChem CID 2829840
IUPAC Name 6-methoxy-1-azatricyclo[7.3.1.0⁵,¹³]trideca-5,7,9(13)-triene
SMILES COC1=C2CCCN3CCCC(C=C1)=C23

Specifications

Quantity 10 g
Formula Weight 203.28
Percent Purity 95%
Chemical Name or Material 8-Methoxyjulolidine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only