accessibility menu, dialog, popup

UserName

H3CON

CAS RN 63468-83-7

IUPAC Name: 6-methoxy-1-azatricyclo[7.3.1.0⁵,¹³]trideca-5,7,9(13)-triene
Synonyms: 6-methoxy-1-azatricyclo[7.3.1.0⁵,¹³]trideca-5,7,9(13)-triene
Molecular Weight (g/mol): 203.29
Molecular Formula: C13H17NO
InChi Key: ZSKPEBROOQBZNW-UHFFFAOYSA-N
SMILES: COC1=C2CCCN3CCCC(C=C1)=C23

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results