missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

8-Hydroxyquinoline (White to Light-Buff Needles or Powder), Fisher BioReagents

Manufacturer:  Fisher BioReagents BP436100

Catalog No. BP436-100




Description

Description

8-Hydroxyquinoline is used to stabilize phenol.
Specifications

Specifications

Hydroxyquinoline
Pass Test
Glass Bottle
4.7hPa at 100°C
White Beige Yellow
100g
≥99.0 %
15, 4881
0.05% max. Insoluble in Alcohol
C1=CC2=C(C(=C1)O)N=CC=C2
145.161
CHEBI:48981
Solid
8-Hydroxyquinoline
0.05% max.
Pass Test (for Mg determination)
267°C
72°C
148-24-3
C9H7NO
8-hydroxyquinoline, 8-oxyquinoline, 8-quinol, 8-quinolinol, bioquin, oxine, oxychinolin, oxyquinoline, phenopyridine, quinophenol
MCJGNVYPOGVAJF-UHFFFAOYSA-N
quinolin-8-ol
1923
145.16
≥99.0%

Chemical Identifiers

148-24-3
145.161
8-hydroxyquinoline, 8-oxyquinoline, 8-quinol, 8-quinolinol, bioquin, oxine, oxychinolin, oxyquinoline, phenopyridine, quinophenol
CHEBI:48981
C1=CC2=C(C(=C1)O)N=CC=C2
C9H7NO
MCJGNVYPOGVAJF-UHFFFAOYSA-N
1923
quinolin-8-ol
Safety and Handling

Safety and Handling

WARNING!

Emergency Overview
Harmful by inhalation, in contact with skin and if swallowed. Possible risks of irreversible effects. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Immediate medical attention is required. Do not induce vomiting.

NFPA

Health:2
Flammability:1
Instability:1

Recommended Storage : RT

SDS Documents
Promotions

Promotions