missing translation for 'onlineSavingsMsg'
Learn More

8-Hydroxyquinoline, ACS

Catalog No. AA41272A1
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA41272A1 1 kg
AA4127218 50 g
AA4127230 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA41272A1 Supplier Thermo Scientific Chemicals Supplier No. 041272A1

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A bacteriostatic and fungistatic agent, also a chelating agent for metals | CAS: 148-24-3 | C9H7NO | 145.16 g/mol

A bacteriostatic and fungistatic agent. Also a chelating agent for metals.8-Hydroxyquinoline is used as an antiseptic and disinfectant. Its alcoholic solution is used in liquid bandages. It is used to prepare tris(8-hydroxyquinolinato)aluminum, which finds application as a component of organic light-emitting diodes (OLED's). It is also used as a reagent and as a stabilizer of hydrogen peroxide in a rocket fuel oxidizer. It is used as a chelating agent and used as a carrier for radio-indium for diagnostic purposes. Its halogenated derivatives are used as anti-infective agents and oral antiamebics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 148-24-3
Molecular Formula C9H7NO
Molecular Weight (g/mol) 145.16
MDL Number MFCD00006807
InChI Key MCJGNVYPOGVAJF-UHFFFAOYSA-N
Synonym 8-hydroxyquinoline, 8-quinolinol, oxyquinoline, oxine, quinophenol, 8-quinol, 8-oxyquinoline, phenopyridine, oxychinolin, bioquin
PubChem CID 1923
ChEBI CHEBI:48981
IUPAC Name quinolin-8-ol
SMILES OC1=C2N=CC=CC2=CC=C1

Specifications

Melting Point 72°C to 76°C
Color White to Yellow
Boiling Point 267°C
Quantity 1 kg
Beilstein 114512
Sensitivity Light sensitive
Merck Index 14,4843
Solubility Information Almost insoluble in water and diethyl ether; Highly soluble in ethanol,acetone,chloroform,and benzene;Soluble in acetic acid,ethanol,acetone,chloroform,benzene and aqueous mineral acids; Insoluble in water and diethylether.
Formula Weight 145.16
Grade ACS
Physical Form Granules
Chemical Name or Material 8-Hydroxyquinoline
Show More Show Less
Hazard Category H301-H317-H318-H360
Hazard Statement GHS H Statement
H341-H302
Suspected of causing genetic defects.
Harmful if swallowed.
Precautionary Statement P201-P202-P261-P264b-P270-P272-P280g-P281-P301+P310-P302+P352-P305+P351+P338-P308+P313-P310-P330-P333+P313-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 205-711-1
RTECSNumber VC4200000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only