Learn More
8-Hydroxyquinoline, ACS reagent, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 121900025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
IndicatorSpecifications
| 8-Hydroxyquinoline | |
| 148-24-3 | |
| White to Yellow or Beige | |
| 0.05% max. | |
| 99% | |
| C9H7NO | |
| 2.5 kg | |
| 15, 4881 | |
| Solubility in water: insoluble. Other solubilities: soluble in alcohol,acetone,chloroform and,benzene | |
| OC1=C2N=CC=CC2=CC=C1 | |
| 145.16 | |
| CHEBI:48981 | |
| ACS Reagent | |
| Crystalline Powder, Crystals or Flakes |
| 99% | |
| 72.0°C to 74.0°C | |
| 267.0°C (752.0 mmHg) | |
| Authentic | |
| Plastic Bottle | |
| MFCD00006807 | |
| 21, IV, 1135 | |
| 8-hydroxyquinoline, 8-quinolinol, oxyquinoline, oxine, quinophenol, 8-quinol, 8-oxyquinoline, phenopyridine, oxychinolin, bioquin | |
| MCJGNVYPOGVAJF-UHFFFAOYSA-N | |
| quinolin-8-ol | |
| 1923 | |
| 145.16 | |
| 0.05% max. (in alcohol) |
Chemical Identifiers
| 148-24-3 | |
| 145.16 | |
| MCJGNVYPOGVAJF-UHFFFAOYSA-N | |
| 1923 | |
| quinolin-8-ol |
| C9H7NO | |
| MFCD00006807 | |
| 8-hydroxyquinoline, 8-quinolinol, oxyquinoline, oxine, quinophenol, 8-quinol, 8-oxyquinoline, phenopyridine, oxychinolin, bioquin | |
| CHEBI:48981 | |
| OC1=C2N=CC=CC2=CC=C1 |
Safety and Handling
GHS H Statement
Toxic if swallowed.
May cause an allergic skin reaction.
Causes serious eye damage.
May damage fertility or the unborn child.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doc
GHS Signal Word: Danger
EINECSNumber : 205-711-1
RTECSNumber : VC4200000
TSCA : TSCA
RUO – Research Use Only