missing translation for 'onlineSavingsMsg'
Learn More

8-Hydroxyquinoline, ACS reagent, Thermo Scientific Chemicals

Catalog No. AC121900250
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
2.5 kg
Packaging:
Glass bottle
Plastic Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC121900250 25 g Glass bottle
AC121901000 100 g Glass bottle
AC121905000 500 g Glass bottle
AC121900010 1 kg Glass bottle
AC121900025 2.5 kg Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. AC121900250 Supplier Thermo Scientific Chemicals Supplier No. 121900250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator

Chemical Identifiers

CAS 148-24-3
Molecular Formula C9H7NO
Molecular Weight (g/mol) 145.16
MDL Number MFCD00006807
InChI Key MCJGNVYPOGVAJF-UHFFFAOYSA-N
Synonym 8-hydroxyquinoline, 8-quinolinol, oxyquinoline, oxine, quinophenol, 8-quinol, 8-oxyquinoline, phenopyridine, oxychinolin, bioquin
PubChem CID 1923
ChEBI CHEBI:48981
IUPAC Name quinolin-8-ol
SMILES OC1=C2N=CC=CC2=CC=C1

Specifications

Melting Point 72.0°C to 74.0°C
Color Beige or White to Yellow
Boiling Point 267.0°C (752.0 mmHg)
Ignition Residue 0.05% max.
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 25 g
Beilstein 21, IV, 1135
Merck Index 15, 4881
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,acetone,chloroform and,benzene
Formula Weight 145.16
Grade ACS Reagent
Insoluble Matter 0.05% max. (in alcohol)
Physical Form Crystalline Powder, Crystals or Flakes
Chemical Name or Material 8-Hydroxyquinoline, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
May cause an allergic skin reaction.
Causes serious eye damage.
May damage fertility or the unborn child.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doc
EINECSNumber 205-711-1
RTECSNumber VC4200000
TSCA TSCA

RUO – Research Use Only