missing translation for 'onlineSavingsMsg'
Learn More

8-Hydroxy-7-iodo-5-quinolinesulfonic Acid, For Spectrophotometric Det. of Fe(III), 98.5%, MilliporeSigma™ Supelco™

Catalog No. p-7193440
Change view
Click to view available options
Quantity:
100 g
Packaging:
Glass Bottle
Catalog No. Quantity Packaging
111018796 100 g Glass Bottle
1 options

Catalog No. 111018796

Supplier: Merck Emd Millipore 55370100G

Only null left

Reagent for the extraction and spectrophotometric determination of Fe

Chemical Identifiers

CAS 547-91-1
Molecular Formula C10H7INNaO7S
Molecular Weight (g/mol) 435.12
MDL Number MFCD00006793
InChI Key FNXKBSAUKFCXIK-UHFFFAOYSA-M
Synonym Ferron; Iodoxyquinolinesulfonic acid
IUPAC Name sodium 8-hydroxy-7-iodoquinoline-5-sulfonic acid hydrogen carbonate
SMILES [Na+].OC([O-])=O.OC1=C2N=CC=CC2=C(C=C1I)S(O)(=O)=O

Specifications

Melting Point 269°C to 270°C (decomposition)
Packaging Glass Bottle
Linear Formula C9H6INO4S
Quantity 100 g
Formula Weight 351.12
Percent Purity ≥98.5%
RTECSNumber VC2800000