missing translation for 'onlineSavingsMsg'
Learn More

8-Hexadecyne 99.0+%, TCI America™

Catalog No. H04345ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H04345ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H04345ML Supplier TCI America Supplier No. H04345ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 19781-86-3
Molecular Formula C16H30
Molecular Weight (g/mol) 222.416
MDL Number MFCD00041664
InChI Key YSDKMBBHKZXPME-UHFFFAOYSA-N
PubChem CID 123387
IUPAC Name hexadec-8-yne
SMILES CCCCCCCC#CCCCCCCC

Specifications

Color Yellow
Quantity 5 mL
Formula Weight 222.42
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 8-Hexadecyne