accessibility menu, dialog, popup

UserName

H3CCH3

CAS RN 19781-86-3

IUPAC Name: hexadec-8-yne
Synonyms: hexadec-8-yne
Molecular Weight (g/mol): 222.42
Molecular Formula: C16H30
InChi Key: YSDKMBBHKZXPME-UHFFFAOYSA-N
SMILES: CCCCCCCC#CCCCCCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results