Learn More
8-Ethoxyquinoline-5-sulfonic acid monohydrate, 99%
CAS: 15301-40-3 | C11H11NO4S | 253.27 g/mol
$83.68 - $1,269.26
Chemical Identifiers
| CAS | 15301-40-3 |
|---|---|
| Molecular Formula | C11H11NO4S |
| Molecular Weight (g/mol) | 253.27 |
| MDL Number | MFCD00065059 |
| InChI Key | YAMVZYRZAMBCED-UHFFFAOYSA-N |
| Synonym | actinoquinol, 8-ethoxy-5-quinolinesulfonic acid, 5-quinolinesulfonic acid, 8-ethoxy, unii-470vqe23o3, 8-ethoxy-5-chinolinsulfonsaeure, actinochinolum, actinoquinolum, actinoquinol inn, actinoquinolum inn-latin, sodium tequinol |
| PubChem CID | 23674 |
| IUPAC Name | 8-ethoxyquinoline-5-sulfonic acid |
| SMILES | CCOC1=C2N=CC=CC2=C(C=C1)S(O)(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1862209
|
Thermo Scientific Chemicals
A1862209 |
10 g |
N/A
|
|
|||||
|
AAA1862218
|
Thermo Scientific Chemicals
A1862218 |
50 g |
N/A
|
|
|||||
|
AAA1862230
|
Thermo Scientific Chemicals
A1862230 |
250 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 15301-40-3 | |
| 253.27 | |
| YAMVZYRZAMBCED-UHFFFAOYSA-N | |
| 23674 | |
| CCOC1=C2N=CC=CC2=C(C=C1)S(O)(=O)=O |
| C11H11NO4S | |
| MFCD00065059 | |
| actinoquinol, 8-ethoxy-5-quinolinesulfonic acid, 5-quinolinesulfonic acid, 8-ethoxy, unii-470vqe23o3, 8-ethoxy-5-chinolinsulfonsaeure, actinochinolum, actinoquinolum, actinoquinol inn, actinoquinolum inn-latin, sodium tequinol | |
| 8-ethoxyquinoline-5-sulfonic acid |
Specifications
| 15301-40-3 | |
| C11H11NO4S | |
| 10 g | |
| actinoquinol, 8-ethoxy-5-quinolinesulfonic acid, 5-quinolinesulfonic acid, 8-ethoxy, unii-470vqe23o3, 8-ethoxy-5-chinolinsulfonsaeure, actinochinolum, actinoquinolum, actinoquinol inn, actinoquinolum inn-latin, sodium tequinol | |
| CCOC1=C2N=CC=CC2=C(C=C1)S(O)(=O)=O | |
| 253.27 | |
| 271.29 (253.28 Anhydrous) | |
| 8-Ethoxyquinoline-5-sulfonic acid monohydrate |
| ∼296°C (decomposition) | |
| MFCD00065059 | |
| 225878 | |
| YAMVZYRZAMBCED-UHFFFAOYSA-N | |
| 8-ethoxyquinoline-5-sulfonic acid | |
| 23674 | |
| 99% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 239-334-9
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only