missing translation for 'onlineSavingsMsg'
Learn More

8-chloronaphthalene-1-sulfonic acid, 97%, Thermo Scientific™

Catalog No. p-2703578
Change view
Click to view available options
Quantity:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BTB06262EA 10g
BTB06262EB 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BTB06262EA Supplier Thermo Scientific Chemicals Supplier No. BTB06262EA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 145-74-4
Molecular Formula C10H7ClO3S
Molecular Weight (g/mol) 242.673
MDL Number MFCD00052379
InChI Key OPFNCYFEIBUZHU-UHFFFAOYSA-N
Synonym 8-chloronaphthalene-1-sulphonic acid, 8-chloro-1-naphthalenesulfonic acid, 1-naphthalenesulfonicacid, 8-chloro, sodium 8-chloronaphthalene-1-sulphonate, maybridge1_001443, 1-naphthalenesulfonic acid, 8-chloro
PubChem CID 67353
IUPAC Name 8-chloronaphthalene-1-sulfonic acid
SMILES C1=CC2=C(C(=C1)S(=O)(=O)O)C(=CC=C2)Cl

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 10g
Formula Weight 242.68
Percent Purity 97%
Chemical Name or Material 8-chloronaphthalene-1-sulfonic acid
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.