missing translation for 'onlineSavingsMsg'
Learn More

8-chloronaphthalene-1-sulfonic acid, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals BTB06262EB

Quantity 25g

 View more versions of this product

Catalog No. BTB06262EB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

145-74-4
242.673
OPFNCYFEIBUZHU-UHFFFAOYSA-N
67353
C1=CC2=C(C(=C1)S(=O)(=O)O)C(=CC=C2)Cl
C10H7ClO3S
MFCD00052379
8-chloronaphthalene-1-sulphonic acid, 8-chloro-1-naphthalenesulfonic acid, 1-naphthalenesulfonicacid, 8-chloro, sodium 8-chloronaphthalene-1-sulphonate, maybridge1_001443, 1-naphthalenesulfonic acid, 8-chloro
8-chloronaphthalene-1-sulfonic acid

Specifications

8-chloronaphthalene-1-sulfonic acid
96.5
97%
25g
8-chloronaphthalene-1-sulphonic acid, 8-chloro-1-naphthalenesulfonic acid, 1-naphthalenesulfonicacid, 8-chloro, sodium 8-chloronaphthalene-1-sulphonate, maybridge1_001443, 1-naphthalenesulfonic acid, 8-chloro
C1=CC2=C(C(=C1)S(=O)(=O)O)C(=CC=C2)Cl
242.673
242.68
145-74-4
100.0
C10H7ClO3S
MFCD00052379
OPFNCYFEIBUZHU-UHFFFAOYSA-N
8-chloronaphthalene-1-sulfonic acid
67353
97%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents