missing translation for 'onlineSavingsMsg'
Learn More

8-(chloromethyl)-6-fluoro-4H-1,3-benzodioxine, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CD08167DA

 View more versions of this product

Catalog No. CD08167DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

8-(Chloromethyl)-6-fluoro-4H-1, 3-benzodioxine
96.5
97%
C9H8ClFO2
1g
FMONGDHUPLQOCP-UHFFFAOYSA-N
8-(chloromethyl)-6-fluoro-4H-1,3-benzodioxine
2780137
97%
131728-94-4
100.0
Amber glass bottle
MFCD00052603
8-chloromethyl-6-fluoro-4h-1,3-benzodioxine, 8-chloromethyl-6-fluoro-4h-benzo d 1,3 dioxine, 4h-1,3-benzodioxin,8-chloromethyl-6-fluoro, 8-chloromethyl-6-fluoro-2,4-dihydro-1,3-benzodioxine, 8-chloromethyl-6-fluorobenzo-1,3-dioxane, acmc-1c5x1, 8-chloromethyl-6-fluoro-4h-benzo 1,3 dioxine, 8-chloromethyl-6-fluoro-2h,4h-1,3-benzodioxine, 8-chloromethyl-6-fluoro-2h,4h-benzo e 1,3-dioxin
FC1=CC(CCl)=C2OCOCC2=C1
202.61
202.61

Chemical Identifiers

131728-94-4
202.61
FMONGDHUPLQOCP-UHFFFAOYSA-N
2780137
FC1=CC(CCl)=C2OCOCC2=C1
C9H8ClFO2
MFCD00052603
8-chloromethyl-6-fluoro-4h-1,3-benzodioxine, 8-chloromethyl-6-fluoro-4h-benzo d 1,3 dioxine, 4h-1,3-benzodioxin,8-chloromethyl-6-fluoro, 8-chloromethyl-6-fluoro-2,4-dihydro-1,3-benzodioxine, 8-chloromethyl-6-fluorobenzo-1,3-dioxane, acmc-1c5x1, 8-chloromethyl-6-fluoro-4h-benzo 1,3 dioxine, 8-chloromethyl-6-fluoro-2h,4h-1,3-benzodioxine, 8-chloromethyl-6-fluoro-2h,4h-benzo e 1,3-dioxin
8-(chloromethyl)-6-fluoro-4H-1,3-benzodioxine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents

Documents

Promotions

Promotions