missing translation for 'onlineSavingsMsg'
Learn More

8-Bromotheophylline 98.0+%, TCI America™

Catalog No. p-7122957
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B34561G 1 g
B34565G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B34561G Supplier TCI America Supplier No. B34561G
Only null left

Chemical Identifiers

CAS 10381-75-6
Molecular Formula C7H7BrN4O2
Molecular Weight (g/mol) 259.063
MDL Number MFCD00022664
InChI Key SKTFQHRVFFOHTQ-UHFFFAOYSA-N
PubChem CID 11808
IUPAC Name 8-bromo-1,3-dimethyl-7H-purine-2,6-dione
SMILES CN1C2=C(C(=O)N(C1=O)C)NC(=N2)Br

Specifications

Melting Point 295°C
Color White-Yellow
Quantity 1 g
Formula Weight 259.06
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 8-Bromotheophylline