accessibility menu, dialog, popup

UserName

CH3NONCH3NBrHNO

CAS RN 10381-75-6

IUPAC Name: 8-bromo-1,3-dimethyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione
Synonyms: pamabrom
Molecular Weight (g/mol): 259.06
Molecular Formula: C7H7BrN4O2
InChi Key: SKTFQHRVFFOHTQ-UHFFFAOYSA-N
SMILES: CN1C2=C(NC(Br)=N2)C(=O)N(C)C1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results