missing translation for 'onlineSavingsMsg'
Learn More
Learn More
8-Bromotheophylline 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 295°C |
| Color | White-Yellow |
| Quantity | 5 g |
| Formula Weight | 259.06 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 8-Bromotheophylline |
Safety and Handling
| TSCA | Yes |