missing translation for 'onlineSavingsMsg'
Learn More

8-Bromoquinoline, 98%, Thermo Scientific Chemicals

Catalog No. AAH2596103
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2596103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2596103 Supplier Thermo Scientific Chemicals Supplier No. H2596103
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 16567-18-3 | Molecular Formula: C9H6BrN | Molecular Weight: 208.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16567-18-3
Molecular Formula C9H6BrN
Molecular Weight (g/mol) 208.06
MDL Number MFCD00191859
InChI Key PIWNKSHCLTZKSZ-UHFFFAOYSA-N
Synonym quinoline, 8-bromo, 8-bromo-quinoline, 8-bromoquinoline, pubchem5816, 8-bromquinoline, acmc-209dt3, ksc181q5t, buttpark 33\04-57
PubChem CID 140109
IUPAC Name 8-bromoquinoline
SMILES BrC1=C2N=CC=CC2=CC=C1

Specifications

Density 1.594
Boiling Point 302°C to 304°C
Flash Point >110°C (230°F)
Refractive Index 1.6725
Quantity 1 g
Sensitivity Light sensitive
Formula Weight 208.06
Percent Purity 98%
Chemical Name or Material 8-Bromoquinoline
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only