accessibility menu, dialog, popup

UserName

CAS RN 16567-18-3

IUPAC Name: 8-bromoquinoline
Synonyms: 8-bromoquinoline
Molecular Weight (g/mol): 208.06
Molecular Formula: C9H6BrN
InChi Key: PIWNKSHCLTZKSZ-UHFFFAOYSA-N
SMILES: BrC1=C2N=CC=CC2=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results