missing translation for 'onlineSavingsMsg'
Learn More

8-Bromoguanosine

Catalog No. AAJ6394006
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6394006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6394006 Supplier Thermo Scientific Chemicals Supplier No. J6394006

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4016-63-1 | C10H12BrN5O5 | 362.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4016-63-1
Molecular Formula C10H12BrN5O5
Molecular Weight (g/mol) 362.14
MDL Number MFCD00037985
InChI Key ASUCSHXLTWZYBA-JJJZDVOVNA-N
Synonym 8-bromoguanosine, 8-bromo-guanosine, guanosine, 8-bromo, 2-amino-8-bromo-9-2r,3r,4s,5r-3,4-dihydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-1h-purin-6 9h-one, 8 brguo, 2-amino-8-bromo-9-2r,3r,4s,5r-3,4-dihydroxy-5-hydroxymethyl oxolan-2-yl-3h-purin-6-one, 8-bro-moguanosine, 8-bromo guanosine, 2-amino-8-bromo-9-2r,3r,4s,5r-3,4-dihydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-1,9-dihydro-6h-purin-6-one, 2-amino-8-bromo-9-2r,3r,4s,5r-3,4-dihydroxy-5-hydroxymethyl oxolan-2-yl-1h-purin-6-one
PubChem CID 92977
IUPAC Name 2-amino-8-bromo-9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one
SMILES NC1=NC2=C(N=C(Br)N2[C@@H]2O[C@H](CO)[C@@H](O)[C@H]2O)C(=O)N1

Specifications

Color White
Quantity 5 g
Formula Weight 362.14
Physical Form Powder
Chemical Name or Material 8-Bromoguanosine
EINECSNumber 223-677-6
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only