missing translation for 'onlineSavingsMsg'
Learn More

8-Bromo-2-methylquinoline 98.0+%, TCI America™

Catalog No. p-7122950
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B34441G 1 g
B34445G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B34441G Supplier TCI America Supplier No. B34441G
Only null left

Chemical Identifiers

CAS 61047-43-6
Molecular Formula C10H8BrN
Molecular Weight (g/mol) 222.09
MDL Number MFCD04966997
InChI Key GQPRZSFQSOEDNV-UHFFFAOYSA-N
Synonym 8-Bromoquinaldine
PubChem CID 4715030
IUPAC Name 8-bromo-2-methylquinoline
SMILES CC1=CC=C2C=CC=C(Br)C2=N1

Specifications

Melting Point 68°C
Color White-Yellow
Boiling Point 132°C
Quantity 1 g
Formula Weight 222.09
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 8-Bromo-2-methylquinoline