accessibility menu, dialog, popup

UserName

H3CNBr

CAS RN 61047-43-6

IUPAC Name: 8-bromo-2-methylquinoline
Synonyms: 8-bromo-2-methylquinoline
Molecular Weight (g/mol): 222.09
Molecular Formula: C10H8BrN
InChi Key: GQPRZSFQSOEDNV-UHFFFAOYSA-N
SMILES: CC1=CC=C2C=CC=C(Br)C2=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results