missing translation for 'onlineSavingsMsg'
Learn More
Learn More
8-Bromo-2-methylquinoline 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 68°C |
| Color | White-Yellow |
| Boiling Point | 132°C |
| Quantity | 1 g |
| Formula Weight | 222.09 |
| Percent Purity | ≥98.0% (GC,T) |
| Grade | Reagent |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 8-Bromo-2-methylquinoline |
Safety and Handling
| TSCA | No |