missing translation for 'onlineSavingsMsg'
Learn More

8-Bromo-1-naphthoic acid, Technical Grade, Maybridge™

$146.81 - $146.81

Chemical Identifiers

CAS 1729-99-3
Molecular Formula C11H7BrO2
Molecular Weight (g/mol) 251.079
InChI Key DMEZDDHJCUHENA-UHFFFAOYSA-N
Synonym 8-bromo-1-naphthoic acid, 1-naphthalenecarboxylic acid, 8-bromo, maybridge1_002138, 8-bromo-1-carboxynaphthalene, 8-bromo-1-naphthalenecarboxylic acid, 1-naphthalenecarboxylicacid, 8-bromo, 8-bromo-naphthalene-1-carboxylic acid
PubChem CID 72873
IUPAC Name 8-bromonaphthalene-1-carboxylic acid
SMILES C1=CC2=C(C(=C1)C(=O)O)C(=CC=C2)Br
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
BTB12326DA
SDS View Documents
Thermo Scientific Chemicals
BTB12326DA
1 g Amber Glass Bottle N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

1729-99-3
251.079
8-bromo-1-naphthoic acid, 1-naphthalenecarboxylic acid, 8-bromo, maybridge1_002138, 8-bromo-1-carboxynaphthalene, 8-bromo-1-naphthalenecarboxylic acid, 1-naphthalenecarboxylicacid, 8-bromo, 8-bromo-naphthalene-1-carboxylic acid
8-bromonaphthalene-1-carboxylic acid
C11H7BrO2
DMEZDDHJCUHENA-UHFFFAOYSA-N
72873
C1=CC2=C(C(=C1)C(=O)O)C(=CC=C2)Br

Specifications

1729-99-3
Amber Glass Bottle
1 g
DMEZDDHJCUHENA-UHFFFAOYSA-N
8-bromonaphthalene-1-carboxylic acid
72873
95%
95%
C11H7BrO2
8-bromo-1-naphthoic acid, 1-naphthalenecarboxylic acid, 8-bromo, maybridge1_002138, 8-bromo-1-carboxynaphthalene, 8-bromo-1-naphthalenecarboxylic acid, 1-naphthalenecarboxylicacid, 8-bromo, 8-bromo-naphthalene-1-carboxylic acid
C1=CC2=C(C(=C1)C(=O)O)C(=CC=C2)Br
251.079
251.08
8-Bromo-1-naphthoic acid
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'